MilliporeSigma
  • Effect of Chitosan on Membrane Permeability of Suspension-Cultured Glycine max and Phaseolus vulgaris Cells.
Effect of Chitosan on Membrane Permeability of Suspension-Cultured Glycine max and Phaseolus vulgaris Cells.
Plant physiology (1982-11-01)
D H Young, H Köhle, H Kauss
ABSTRACT
Treatment of suspension-cultured Glycine max cv Harosoy 63 cells with soluble chitosan (20-500 micrograms per milliliter) increased membrane permeability as shown by leakage of electrolytes, protein, and UV absorbing material. Severe damage to the cell membrane by chitosan (100 and 500 mug/ml) was also indicated by reduced staining with fluorescein diacetate and the leakage of fluorescein from preloaded cells. Other basic polymers (poly-l-lysine, histone, DEAE-dextran, protamine sulfate, and glycol chitosan) also increased permeability, whereas the basic monomers l-lysine and d-glucosamine, and acidic or neutral polymers were not active. Chitosan-induced leakage was inhibited by divalent cations, the order of effectiveness being Ba(2+) > Ca(2+) > Sr(2+) > Mg(2+). Na polygalacturonate and Na poly-l-aspartate also reduced polycation-induced leakage, probably by formation of polycation-polyanion complexes. A chitosan-polygalacturonate complex precipitated on mixing solutions of the two polymers containing approximately equal numbers of galacturonate and glucosamine residues, but not with either polymer in excess. A similar concentration-dependent precipitation of chitosan by Na poly-l-aspartate was found. Leakage from Phaseolus vulgaris cv Grandessa cells was also induced by chitosan, and was inhibited by Ca(2+) and Na polygalacturonate.
Sigma-Aldrich
Glycol chitosan, ≥60% (titration), crystalline
Pricing and availability is not currently available.