MilliporeSigma
Efficient enzymatic hydrolysis of polyprenyl pyrophosphates.
Biochimica et biophysica acta (1982-09-14)
H Fujii, T Koyama, K Ogura
ABSTRACT
A method of efficient enzymatic hydrolysis of polyprenyl pyrophosphates was established. Polyprenyl pyrophosphates such as all-trans-octaprenyl and all-trans-nonaprenyl (solanesyl) pyrophosphate can be hydrolyzed completely to the corresponding primary alcohols by the action of potato acid phosphatase in the presence of 60% (v/v) methanol.