MilliporeSigma
  • Lipopolysaccharide is a frequent contaminant of plasmid DNA preparations and can be toxic to primary human cells in the presence of adenovirus.
Lipopolysaccharide is a frequent contaminant of plasmid DNA preparations and can be toxic to primary human cells in the presence of adenovirus.
Gene therapy (1994-07-01)
M Cotten, A Baker, M Saltik, E Wagner, M Buschle
ABSTRACT
Endotoxin (lipopolysaccharide, LPS) is commonly found as a contaminant in plasmid DNA preparations. We demonstrate here that the quantities of LPS typically contaminating DNA preparations can generate a toxicity to primary cells (primary human skin fibroblasts, primary human melanoma cells) in the presence of entry-competent adenovirus particles. Toxicity can be observed with as little as 100 ng/ml free LPS or 100 pg/ml LPS when the LPS is assembled into polylysine/adenovirus complexes. Simple and effective methods of removing the contaminating LPS using either a polymyxin B resin or Triton X-114 extraction are described. Treatment of DNA samples to remove LPS eliminates the toxicity to primary cells.
Sigma-Aldrich
Endotoxin Removal solution, Remove lipopolysaccharides (LPS) from plasmid DNA preparations
Sigma-Aldrich
Triton X-114, laboratory grade