MilliporeSigma
  • Calcium phosphate microcrystals in the renal tubular fluid accelerate chronic kidney disease progression.
Calcium phosphate microcrystals in the renal tubular fluid accelerate chronic kidney disease progression.
The Journal of clinical investigation (2021-06-30)
Kazuhiro Shiizaki, Asako Tsubouchi, Yutaka Miura, Kinya Seo, Takahiro Kuchimaru, Hirosaka Hayashi, Yoshitaka Iwazu, Marina Miura, Batpurev Battulga, Nobuhiko Ohno, Toru Hara, Rina Kunishige, Mamiko Masutani, Keita Negishi, Kazuomi Kario, Kazuhiko Kotani, Toshiyuki Yamada, Daisuke Nagata, Issei Komuro, Hiroshi Itoh, Hiroshi Kurosu, Masayuki Murata, Makoto Kuro-O
ABSTRACT
The Western pattern diet is rich not only in fat and calories but also in phosphate. The negative effects of excessive fat and calorie intake on health are widely known, but the potential harms of excessive phosphate intake are poorly recognized. Here, we show the mechanism by which dietary phosphate damages the kidney. When phosphate intake was excessive relative to the number of functioning nephrons, circulating levels of FGF23, a hormone that increases the excretion of phosphate per nephron, were increased to maintain phosphate homeostasis. FGF23 suppressed phosphate reabsorption in renal tubules and thus raised the phosphate concentration in the tubule fluid. Once it exceeded a threshold, microscopic particles containing calcium phosphate crystals appeared in the tubule lumen, which damaged tubule cells through binding to the TLR4 expressed on them. Persistent tubule damage induced interstitial fibrosis, reduced the number of nephrons, and further boosted FGF23 to trigger a deterioration spiral leading to progressive nephron loss. In humans, the progression of chronic kidney disease (CKD) ensued when serum FGF23 levels exceeded 53 pg/mL. The present study identified calcium phosphate particles in the renal tubular fluid as an effective therapeutic target to decelerate nephron loss during the course of aging and CKD progression.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, ~2.0 mg/mL, clone AA2, purified from hybridoma cell culture
Sigma-Aldrich
TLR4 Inhibitor, TAK-242, TAK-242; CAS 243984-11-4; is a cell-permeable compound that selectively binds to Cys747 of TLR4 and selectively disrupts its interaction with adaptor molecules TIRAP and TRAM.