MilliporeSigma
  • Evaluation of ELISA-Based Multiplex Peptides for the Detection of Human Serum Antibodies Induced by Zika Virus Infection across Various Countries.
Evaluation of ELISA-Based Multiplex Peptides for the Detection of Human Serum Antibodies Induced by Zika Virus Infection across Various Countries.
Viruses (2021-08-11)
Maria Del Pilar Martinez Viedma, Stephen Panossian, Kennedy Gifford, Kimberly García, Isis Figueroa, Leda Parham, Laise de Moraes, Lillian Nunes Gomes, Tamara García-Salum, Cecilia Perret, Daniela Weiskopf, Gene S Tan, Antônio Augusto Silva, Viviane Boaventura, Guillermo M Ruiz-Palacios, Alessandro Sette, Aruna Dharshan De Silva, Rafael A Medina, Ivette Lorenzana, Kevan M Akrami, Ricardo Khouri, Daniel Olson, Brett E Pickett
ABSTRACT
Zika virus (ZIKV) is a mosquito-borne Flavivirus with a positive-sense RNA genome, which are generally transmitted through the bite of an infected Aedes mosquito. ZIKV infections could be associated with neurological sequelae that, and otherwise produces similar clinical symptoms as other co-circulating pathogens. Past infection with one member of the Flavivirus genus often induces cross-reactive antibodies against other flaviruses. These attributes complicate the ability to differentially diagnose ZIKV infection from other endemic mosquito-borne viruses, making it both a public health issue as well as a diagnostic challenge. We report the results from serological analyses using arbovirus-specific peptides on 339 samples that were previously collected from 6 countries. Overall, we found that our multiplexed peptide-based ELISA was highly efficient for identifying ZIKV antibodies as early as 2 weeks post infection, and that it correlates with microneutralization, plaque reduction neutralization tests (PRNTs) and commercial tests for ZIKV in previously characterized samples. We observed that seropositivity varied by patient cohort, reflecting the sampling period in relation to the 2015-2016 ZIKV outbreak. This work evaluates the accuracy, specificity, and sensitivity of our peptide-based ELISA method for detecting ZIKV antibodies from geographically diverse regions. These findings can contribute to ongoing serological methods development and can be adapted for use in future studies.
Sigma-Aldrich
Complement component C1q from human serum, ≥95% (SDS-PAGE)