MilliporeSigma
  • Screening of metallic pollution in complex environmental samples through a transcriptomic fingerprint method.
Screening of metallic pollution in complex environmental samples through a transcriptomic fingerprint method.
Environmental science and pollution research international (2021-08-04)
Mickael Cregut, Anna Hua, Sulivan Jouanneau, Ali Assaf, Christophe B Y Cordella, Gérald Thouand, Marie-José Durand
ABSTRACT
Characterizing waste ecotoxicity is laborious because of both the undefined nature of environmental samples and the diversity of contaminants that can be present. With regard to these limitations, traditional approaches do not provide information about the nature of the pollution encountered. To improve such assessments, a fluorescent library of 1870 transcriptomic reporters from Escherichia coli K12 MG1655 was used to report the ecotoxic status of environmental samples. The reliability of the approach was evaluated with 6 metallic pollutants (As, Cu, Cd, Hg, Pb, Zn) used alone and in mixture in pure and complex matrices. A total of 18 synthetic samples were used to characterize the specificity of the resulting metallic contamination fingerprints. Metallic contamination impacted 4.5 to 10.2% of the whole transcriptomic fingerprint of E. coli. The analysis revealed that a subset of 175 transcriptomic reporters is sufficient to characterize metallic contamination, regardless of the nature of the sample. A statistical model distinguished patterns due to metallic contamination and provided information about the level of toxicity with 93 to 98% confidence. The use of the transcriptomic assessment was validated for 17 complex matrices with various toxicities and metal contaminants, such as activated sludge, wastewater effluent, soil, wood and river water. The presence of metals and their associated toxicity, which seems linked to their bioavailabilities, were thereby determined. This method constitutes a possible tool to screen unknown complex samples for their metallic status and identify those for which a deeper characterization must be achieved by the use of traditional biosensors and analytical methods.
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Anti-TGIF2 Antibody, Upstate®, from rabbit
Sigma-Aldrich
D-(+)-Glucose monohydrate, meets analytical specification of Ph. Eur., BP, Ph Franç., 97.5-102.0% anhydrous basis(HPLC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)