MilliporeSigma
  • Regulation of Chemosensitivity in Human Medulloblastoma Cells by p53 and the PI3 Kinase Signaling Pathway.
Regulation of Chemosensitivity in Human Medulloblastoma Cells by p53 and the PI3 Kinase Signaling Pathway.
Molecular cancer research : MCR (2021-10-13)
Aisha Naeem, Varsha Harish, Sophie Coste, Erika M Parasido, Muhammad Umer Choudhry, Lawrence F Kromer, Chukuemeka Ihemelandu, Emanuel F Petricoin, Mariaelena Pierobon, Muhammad Saad Noon, Venkata Mahidhar Yenugonda, Maria Avantaggiati, Gary M Kupfer, Stanley Fricke, Olga Rodriguez, Chris Albanese
ABSTRACT
In medulloblastoma, p53 expression has been associated with chemoresistance and radiation resistance and with poor long-term outcomes in the p53-mutated sonic hedgehog, MYC-p53, and p53-positive medulloblastoma subgroups. We previously established a direct role for p53 in supporting drug resistance in medulloblastoma cells with high basal protein expression levels (D556 and DAOY). We now show that p53 genetic suppression in medulloblastoma cells with low basal p53 protein expression levels (D283 and UW228) significantly reduced drug responsiveness, suggesting opposing roles for low p53 protein expression levels. Mechanistically, the enhanced cell death by p53 knockdown in high-p53 cells was associated with an induction of mTOR/PI3K signaling. Both mTOR inhibition and p110α/PIK3CA induction confirmed these findings, which abrogated or accentuated the enhanced chemosensitivity response in D556 cells respectively while converse was seen in D283 cells. Co-treatment with G-actin-sequestering peptide, thymosin β4 (Tβ4), induced p-AKTS473 in both p53-high and p53-low cells, enhancing chemosensitivity in D556 cells while enhancing chemoresistance in D283 and UW228 cells. IMPLICATIONS: Collectively, we identified an unexpected role for the PI3K signaling in enhancing cell death in medulloblastoma cells with high basal p53 expression. These studies indicate that levels of p53 immunopositivity may serve as a diagnostic marker of chemotherapy resistance and for defining therapeutic targeting.
Sigma-Aldrich
Anti-p53 Antibody, clone BP53-12, clone BP53-12, Upstate®, from mouse
Sigma-Aldrich
Everolimus, ≥95% (HPLC)
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications