MilliporeSigma
  • C1r Upregulates Production of Matrix Metalloproteinase-13 and Promotes Invasion of Cutaneous Squamous Cell Carcinoma.
C1r Upregulates Production of Matrix Metalloproteinase-13 and Promotes Invasion of Cutaneous Squamous Cell Carcinoma.
The Journal of investigative dermatology (2021-11-11)
Kristina Viiklepp, Liisa Nissinen, Marjaana Ojalill, Pilvi Riihilä, Markku Kallajoki, Seppo Meri, Jyrki Heino, Veli-Matti Kähäri
ABSTRACT
Cutaneous squamous cell carcinoma (cSCC) is the most common metastatic skin cancer with increasing incidence worldwide. Previous studies have demonstrated the role of complement system in cSCC progression. In this study we have investigated the mechanistic role of serine protease C1r, a component of the classical pathway of complement system, in cSCC. Knockout of C1r in cSCC cells using CRISPR/Cas9 resulted in significant decrease in their proliferation, migration, and invasion through collagen type I compared to wild type cSCC cells. Knockout of C1r suppressed growth and vascularization of cSCC xenograft tumors, and promoted apoptosis of tumor cells in vivo. mRNA-seq analysis after C1r knockdown revealed significantly regulated GO terms Cell-matrix adhesion, Extracellular matrix component, Basement membrane, Metalloendopeptidase activity and KEGG pathway Extracellular matrix-receptor interaction. Among the significantly regulated genes were invasion-associated matrix metalloproteinases MMP1, MMP13, MMP10, and MMP12. Knockout of C1r resulted in decreased production of MMP-1, MMP-13, MMP-10, and MMP-12 by cSCC cells in culture. Knockout of C1r inhibited expression of MMP-13 by tumor cells, suppressed invasion, and reduced the amount of degraded collagen in vivo in xenografts. These results provide evidence for the role of C1r in promoting the invasion of cSCC cells by increasing MMP production.
Sigma-Aldrich
Anti-C1S antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-C1R antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MMP-13 Inhibitor, The MMP-13 Inhibitor, also referenced under CAS 544678-85-5, controls the biological activity of MMP-13. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
Anti-MMP-13 Antibody, clone VIIIA2, clone VIIIA2, Chemicon®, from mouse