MilliporeSigma
  • UHPLC-MS-based metabolomics and chemoinformatics study reveals the neuroprotective effect and chemical characteristic in Parkinson's disease mice after oral administration of Wen-Shen-Yang-Gan decoction.
UHPLC-MS-based metabolomics and chemoinformatics study reveals the neuroprotective effect and chemical characteristic in Parkinson's disease mice after oral administration of Wen-Shen-Yang-Gan decoction.
Aging (2021-08-03)
Guoxue Zhu, Wang Wang, Chang Chen, Lili Tang, Yan Liang, Zhennian Zhang, Yan Lu, Yang Zhao
ABSTRACT
Parkinson's disease (PD), the typical neurodegenerative disease, is characterized by the progressive loss of dopaminergic neurons in the substantia nigra (SN). However, no therapeutic agent used currently could slow down neuronal cell loss so as to decelerate or halt the progression of PD. Traditional Chinese medicine (TCM) has been utilized to treat the dysfunction of the autonomic nervous system. Wen-Shen-Yang-Gan decoction (WSYGD) has a good effect on the clinical treatment of PD with constipation. However, it is not clear which ingredients and what mechanism are responsible for the therapeutic effect. In this study, the pharmacodynamic study of WSYGD in PD mice was applied. Concurrently, a novel method for the identification of metabolic profiles of WSYGD has been developed. Finally, we found that WSYGD could protect the PD mice induced by rotenone. The underlying mechanism of the protective effect may be related to the reduction of the DA neurons apoptosis via reducing inflammatory reaction. By virtue of UPLC-MS and chemoinformatics method, 35 prototype compounds and 27 metabolites were filtered out and tentatively characterized. In conclusion, this study provides an insight into the metabolism of WSYGD in vivo to enable understanding of the metabolic process and therapeutic mechanism of PD.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Anti-Tyrosine Hydroxylase antibody, Mouse monoclonal, clone TH-16, purified from hybridoma cell culture