MilliporeSigma
  • Delivering Antisense Oligonucleotides across the Blood-Brain Barrier by Tumor Cell-Derived Small Apoptotic Bodies.
Delivering Antisense Oligonucleotides across the Blood-Brain Barrier by Tumor Cell-Derived Small Apoptotic Bodies.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2021-07-15)
Yulian Wang, Jiayun Pang, Qingyun Wang, Luocheng Yan, Lintao Wang, Zhen Xing, Chunming Wang, Junfeng Zhang, Lei Dong
ABSTRACT
The blood-brain barrier (BBB) is the most restrictive and complicated barrier that keeps most biomolecules and drugs from the brain. An efficient brain delivery strategy is urgently needed for the treatment of brain diseases. Based on the studies of brain-targeting extracellular vesicles (EVs), the potential of using small apoptotic bodies (sABs) from brain metastatic cancer cells for brain-targeting drug delivery is explored. It is found that anti-TNF-α antisense oligonucleotide (ASO) combined with cationic konjac glucomannan (cKGM) can be successfully loaded into sABs via a transfection/apoptosis induction process and that the sABs generated by B16F10 cells have an extraordinarily high brain delivery efficiency. Further studies suggest that ASO-loaded sABs (sCABs) are transcytosed by b. End3 (brain microvascular endothelial cells, BMECs) to penetrate the BBB, which is mediated by CD44v6, and eventually taken up by microglial cells in the brain. In a Parkinson's disease (PD) mouse model, sCABs dramatically ameliorate PD symptoms via the anti-inflammatory effect of ASO. This study suggests that sABs from brain metastatic cancer cells are excellent carriers for brain-targeted delivery, as they have not only an extraordinary delivery efficiency but also a much higher scale-up production potential than other EVs.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Anti-CD44v6 Antibody, exon v6, Chemicon®, from rabbit