MilliporeSigma
  • Protein expression of prognostic genes in primary melanoma and benign nevi.
Protein expression of prognostic genes in primary melanoma and benign nevi.
Journal of cancer research and clinical oncology (2021-11-11)
T Gambichler, J Elfering, T Meyer, S Bruckmüller, E Stockfleth, M Skrygan, H U Käfferlein, T Brüning, K Lang, D Wagener, S Schröder, M Nick, L Susok
ABSTRACT
To evaluate the protein expression characteristics of genes employed in a recently introduced prognostic gene expression assay for patients with cutaneous melanoma (CM). We studied 37 patients with CM and 10 with benign (melanocytic) nevi (BN). Immunohistochemistry of primary tumor tissue was performed for eight proteins: COL6A6, DCD, GBP4, KLHL41, KRT9, PIP, SCGB1D2, SCGB2A2. The protein expression of most markers investigated was relatively low (e.g., DCD, KRT9, SCGB1D2) and predominantly cytoplasmatic in melanocytes and keratinocytes. COL6A6, GBP4, and KLHL41 expression was significantly enhanced in CM when compared to BN. DCD protein expression was significantly correlated with COL6A6, GBP4, and KLHL41. GBP4 was positively correlated with KLHL41 and inversely correlated with SCGB2B2. The latter was also inversely correlated with serum S100B levels at time of initial diagnosis. The presence of SCGB1D2 expression was significantly associated with ulceration of the primary tumor. KRT9 protein expression was significantly more likely found in acral lentiginous melanoma. The presence of DCD expression was less likely associated with superficial spreading melanoma subtype but significantly associated with non-progressive disease. The absence of SCGB2A2 expression was significantly more often observed in patients who did not progress to stage III or IV. The expression levels observed were relatively low but differed in part with those found in BN. Even though we detected some significant correlations between the protein expression levels and clinical parameters (e.g., CM subtype, course of disease), there was no major concordance with the protective or risk-associated functions of the corresponding genes included in a recently introduced prognostic gene expression assay.
Sigma-Aldrich
Anti-DCD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Anti-PIP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-KLHL41 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2