MilliporeSigma
  • Age-related changes in the local milieu of inflamed tissues cause aberrant neutrophil trafficking and subsequent remote organ damage.
Age-related changes in the local milieu of inflamed tissues cause aberrant neutrophil trafficking and subsequent remote organ damage.
Immunity (2021-05-26)
Anna Barkaway, Loïc Rolas, Régis Joulia, Jennifer Bodkin, Tchern Lenn, Charlotte Owen-Woods, Natalia Reglero-Real, Monja Stein, Laura Vázquez-Martínez, Tamara Girbl, Robin N Poston, Matthew Golding, Rebecca S Saleeb, Aude Thiriot, Ulrich H von Andrian, Johan Duchene, Mathieu-Benoit Voisin, Cleo L Bishop, David Voehringer, Axel Roers, Antal Rot, Tim Lämmermann, Sussan Nourshargh
ABSTRACT
Aging is associated with dysregulated immune functions. Here, we investigated the impact of age on neutrophil diapedesis. Using confocal intravital microscopy, we found that in aged mice, neutrophils adhered to vascular endothelium in inflamed tissues but exhibited a high frequency of reverse transendothelial migration (rTEM). This retrograde breaching of the endothelium by neutrophils was governed by enhanced production of the chemokine CXCL1 from mast cells that localized at endothelial cell (EC) junctions. Increased EC expression of the atypical chemokine receptor 1 (ACKR1) supported this pro-inflammatory milieu in aged venules. Accumulation of CXCL1 caused desensitization of the chemokine receptor CXCR2 on neutrophils and loss of neutrophil directional motility within EC junctions. Fluorescent tracking revealed that in aged mice, neutrophils undergoing rTEM re-entered the circulation and disseminated to the lungs where they caused vascular leakage. Thus, neutrophils stemming from a local inflammatory site contribute to remote organ damage, with implication to the dysregulated systemic inflammation associated with aging.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Evans Blue, Dye content ≥75 %