MilliporeSigma
  • A rapid, ultrasensitive voltammetric biosensor for determining SARS-CoV-2 spike protein in real samples.
A rapid, ultrasensitive voltammetric biosensor for determining SARS-CoV-2 spike protein in real samples.
Biosensors & bioelectronics (2021-07-19)
Lokman Liv, Gizem Çoban, Nuri Nakiboğlu, Tanıl Kocagöz
ABSTRACT
The ongoing coronavirus disease 2019 (COVID-19) pandemic continues to threaten public health systems all around the world. In controlling the viral outbreak, early diagnosis of COVID-19 is pivotal. This article describes a novel method of voltammetrically determining severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) spike protein with a newly designed sensor involving bovine serum albumin, SARS-CoV-2 spike antibody and a functionalised graphene oxide modified glassy carbon electrode (BSA/AB/f-GO/GCE) or screen-printed electrode (BSA/AB/f-GO/SPE). The oxidation reaction based on the antibody-antigen protein interaction was evaluated as a response to SARS-CoV-2 spike protein at -200 mV and 1430 mV with the BSA/AB/f-GO/SPE and BSA/AB/f-GO/GCE, respectively. The developed sensors, BSA/AB/f-GO/SPE and BSA/AB/f-GO/GCE, could detect 1 ag/mL of virus spike protein in synthetic, saliva and oropharyngeal swab samples in 5 min and 35 min, and both sensors demonstrated a dynamic response to the SARS-CoV-2 spike protein between 1 ag/mL and 10 fg/mL. Real-time polymerase chain reaction (RT-PCR), rapid antigen test and the proposed method were applied to saliva samples. When compared to RT-PCR, it was observed that the developed method had a 92.5% specificity and 93.3% sensitivity. Moreover, BSA/AB/f-GO/SPE sensor achieved 91.7% accuracy compared to 66.7% accuracy of rapid antigen test kit in positive samples. In view of these findings, the developed sensor provides great potential for the diagnosing of COVID-19 in real samples.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, purum, ≥98.0% (AT)