MilliporeSigma
  • Differential Protein Metabolism and Regeneration in Gastrocnemius Muscles in High-fat Diet Fed Mice and Pre-hibernation Daurian Ground Squirrels: A Comparison between Pathological and Healthy Obesity.
Differential Protein Metabolism and Regeneration in Gastrocnemius Muscles in High-fat Diet Fed Mice and Pre-hibernation Daurian Ground Squirrels: A Comparison between Pathological and Healthy Obesity.
Zoological studies (2021-08-14)
Xia Yan, Qiaohua Niu, Xuli Gao, Shenyang Shen, Nan He, Huiping Wang, Rongrong Fang, Yunfang Gao, Hui Chang
ABSTRACT
We focused on pathological obesity induced by excessive fat intake (nutritional obesity) in non-hibernator and healthy obesity due to pre-hibernation (PRE) fat storage in hibernator to study the effects of different types of obesity on skeletal muscle protein metabolism and cell regeneration. Kunming mice were fed with high-fat diet for 3 months to construct a pathological obesity model. Daurian ground squirrels fattened naturally before hibernation were used as a healthy obesity model. Body weight, adipose tissue wet weight, gastrocnemius muscle wet weight, muscle fiber cross-sectional area (CSA) and fiber type distribution were measured. The protein expression levels related to protein degradation (MuRF-1, atrogin-1, calpain1, calpain2, calpastatin, desmin, troponin T, Beclin-1, LC3-II), protein synthesis (P-Akt, P-mTORC1, P-S6K1, P-4E-BP1) and cell regeneration (MyoD, myogenin, myostatin) were detected by Western blot. As a result, the body weight and adipose tissue wet weight were both significantly increased in high fat obese (OB) mice and pre-hibernation fat (PRE) ground squirrels. The muscle wet weight, ratio of muscle wet weight to body weight, and muscle fiber CSA were significantly decreased, while the percentage of MHC I fiber isoform was significantly increased in gastrocnemius muscle of OB mice compared with the control (CON) group. The protein expression levels of P-Akt, P-mTORC1, P-4E-BP1 and myogenin were significantly decreased, while those of calpain1, calpain2, MuRF-1 and myostatin were significantly increased in the OB mice. In the ground squirrels, the muscle wet weight, muscle fiber CSA and percentage of MHC I fiber isoform all showed no change in the gastrocnemius muscle in the PRE group compared with the summer active (SA) group. The protein expression levels of P-Akt, P-mTORC1, P-S6K1 and MyoD were significantly increased, while those of Beclin-1 and LC3-II were significantly decreased in the PRE ground squirrels. This study demonstrated that the decrease in protein expression levels in the Akt/mTOR pathway (P-Akt, P-mTORC1 and P-4E-BP1) and cell regeneration (myogenin) and the increase in protein expression levels of the calpain pathway (calpain1 and calpain2) and ubiquitin-proteasome pathway (MuRF-1) were involved in the mechanism of muscle atrophy in gastrocnemius muscle of the pathologically obese Kunming mice induced by high-fat diet. In contrast, the increased protein expression levels of the Akt/mTOR pathway (P-Akt, P-mTORC1 and P-S6K1) and cell regeneration (MyoD), and the decreased protein expression levels of the autophagy lysosomal pathway (Beclin-1 and LC3-II) were involved in the mechanism of anti-atrophy in gastrocnemius muscle of the healthy obese ground squirrels fattened before hibernation.
Sigma-Aldrich
Anti-Mouse Polyvalent Immunoglobulins (G,A,M)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Troponin T antibody produced in mouse, clone JLT-12, ascites fluid