MilliporeSigma
  • Influence of Algae Supplementation on the Concentration of Glutathione and the Activity of Glutathione Enzymes in the Mice Liver and Kidney.
Influence of Algae Supplementation on the Concentration of Glutathione and the Activity of Glutathione Enzymes in the Mice Liver and Kidney.
Nutrients (2021-07-03)
Grażyna Świderska-Kołacz, Małgorzata Jefimow, Jolanta Klusek, Norbert Rączka, Szymon Zmorzyński, Anna Wojciechowska, Iwona Stanisławska, Marek Łyp, Joanna Czerwik-Marcinkowska
ABSTRACT
Algae are potential and natural source of long-chain polyunsaturated fatty acids like eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA). The diatom Pinnularia borealis accumulates high levels of EPA and may be considered as a source for commercial production of dietary supplements. In this study we asked the question whether diet supplementation with P. borealis may augment antioxidant defense and ameliorate risk factors for cardiovascular diseases. We fed mice (Mus musculus) with lyophilized diatom solutions of different concentrations (1%, 3%, and 5%) for 7 days. Then we measured glutathione content and the activity of glutathione redox system enzymes, total cholesterol and triacylglycerol concentrations, and malondialdehyde concentration in the liver and kidney. We found that cholesterol and triacylglycerol concentrations in the liver and kidneys were the lowest in mice who were fed with the highest concentration of Pinnularia borealis, suggesting protective properties of algae. Additionally, the lowest concentration of Pinnularia borealis was sufficient to improve antioxidant capacity. Our results suggest that P. borealis may be used as a source for dietary supplements rich in EPA, but the amount supplied to the organism should be limited.
Sigma-Aldrich
Glutathione-S-Transferase (GST) Assay Kit, sufficient for 500 multiwell tests
Sigma-Aldrich
Glutathione Assay Kit, sufficient for 700 assays
Sigma-Aldrich
Glutathione Reductase Assay Kit, Sufficient for 100 colorimetric tests