MilliporeSigma
  • A Simple and Efficient Method to Generate Dual Site-Specific Conjugation ADCs with Cysteine Residue and an Unnatural Amino Acid.
A Simple and Efficient Method to Generate Dual Site-Specific Conjugation ADCs with Cysteine Residue and an Unnatural Amino Acid.
Bioconjugate chemistry (2021-05-21)
Lin Zhang, Zewei Wang, Zhiyuan Wang, Fang Luo, Mingfeng Guan, Meimei Xu, Yundong Li, Yaoyang Zhang, Zhaoyin Wang, Wenyuan Wang
ABSTRACT
Antibody-drug conjugates (ADCs) are complex pharmaceutical molecules that combine monoclonal antibodies with biologically active drugs through chemical linkers. ADCs are designed to specifically kill disease cells by utilizing the target specificity of antibodies and the cytotoxicity of chemical drugs. However, the traditional ADCs were only applied to a few disease targets because of some limitations such as the huge molecular weight, the uncontrollable coupling reactions, and a single mechanism of action. Here we report a simple, one-pot, successive reaction method to produce dual payload conjugates with the site-specifically engineered cysteine and p-acetyl-phenylalanine using Herceptin (trastuzumab), an anti-HER2 antibody drug widely used for breast cancer treatment, as a tool molecule. This strategy enables antibodies to conjugate with two mechanistically distinct cytotoxic drugs through different functional groups sequentially, therefore, rendering the newly designed ADCs with functional diversity and the potential to overcome drug resistance and enhance the therapeutic efficacy.
Sigma-Aldrich
Anti-Human Kappa Light Chains (Bound and Free)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution