MilliporeSigma
  • A ligand-insensitive UNC5B splicing isoform regulates angiogenesis by promoting apoptosis.
A ligand-insensitive UNC5B splicing isoform regulates angiogenesis by promoting apoptosis.
Nature communications (2021-08-13)
Davide Pradella, Gianluca Deflorian, Alex Pezzotta, Anna Di Matteo, Elisa Belloni, Daniele Campolungo, Andrea Paradisi, Mattia Bugatti, William Vermi, Matteo Campioni, Antonella Chiapparino, Luigi Scietti, Federico Forneris, Costanza Giampietro, Nina Volf, Michael Rehman, Serena Zacchigna, Maria Paola Paronetto, Anna Pistocchi, Anne Eichmann, Patrick Mehlen, Claudia Ghigna
ABSTRACT
The Netrin-1 receptor UNC5B is an axon guidance regulator that is also expressed in endothelial cells (ECs), where it finely controls developmental and tumor angiogenesis. In the absence of Netrin-1, UNC5B induces apoptosis that is blocked upon Netrin-1 binding. Here, we identify an UNC5B splicing isoform (called UNC5B-Δ8) expressed exclusively by ECs and generated through exon skipping by NOVA2, an alternative splicing factor regulating vascular development. We show that UNC5B-Δ8 is a constitutively pro-apoptotic splicing isoform insensitive to Netrin-1 and required for specific blood vessel development in an apoptosis-dependent manner. Like NOVA2, UNC5B-Δ8 is aberrantly expressed in colon cancer vasculature where its expression correlates with tumor angiogenesis and poor patient outcome. Collectively, our data identify a mechanism controlling UNC5B's necessary apoptotic function in ECs and suggest that the NOVA2/UNC5B circuit represents a post-transcriptional pathway regulating angiogenesis.
Sigma-Aldrich
Anti-phospho-DAP-Kinase (pSer308) antibody, Mouse monoclonal, clone DKPS308, purified from hybridoma cell culture
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
MCDB 131 Medium, With trace elements and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Anti-Green Fluorescent Protein Antibody, Chemicon®, from mouse
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Endothelial cell growth supplement from bovine neural tissue, ECGS, suitable for cell culture
Sigma-Aldrich
Anti-NOVA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Vinculin Antibody, clone VIIF9 (7F9), clone VIIF9 (7F9), Chemicon®, from mouse
Sigma-Aldrich
Anti-DAP Kinase 1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution