MilliporeSigma
Total synthesis of isoroquefortine C.
The Journal of organic chemistry (2002-02-22)
Bruno M Schiavi, David J Richard, Madeleine M Joullié
ABSTRACT
A short and efficient total synthesis of isoroquefortine C, the 3,12-(Z)-isomer of roquefortine C, from L-tryptophan methyl ester hydrochloride and 4(5)-(hydroxy)methylimidazole hydrochloride is described.
Sigma-Aldrich
L-Tryptophan methyl ester hydrochloride, 98%