MilliporeSigma
2-Sulfonylpyridines as Tunable, Cysteine-Reactive Electrophiles.
Journal of the American Chemical Society (2020-04-18)
Claudio Zambaldo, Ekaterina V Vinogradova, Xiaotian Qi, Jonathan Iaconelli, Radu M Suciu, Minseob Koh, Kristine Senkane, Stormi R Chadwick, Brittany B Sanchez, Jason S Chen, Arnab K Chatterjee, Peng Liu, Peter G Schultz, Benjamin F Cravatt, Michael J Bollong
ABSTRACT
The emerging use of covalent ligands as chemical probes and drugs would benefit from an expanded repertoire of cysteine-reactive electrophiles for efficient and diverse targeting of the proteome. Here we use the endogenous electrophile sensor of mammalian cells, the KEAP1-NRF2 pathway, to discover cysteine-reactive electrophilic fragments from a reporter-based screen for NRF2 activation. This strategy identified a series of 2-sulfonylpyridines that selectively react with biological thiols via nucleophilic aromatic substitution (SNAr). By tuning the electrophilicity and appended recognition elements, we demonstrate the potential of the 2-sulfonylpyridine reactive group with the discovery of a selective covalent modifier of adenosine deaminase (ADA). Targeting a cysteine distal to the active site, this molecule attenuates the enzymatic activity of ADA and inhibits proliferation of lymphocytic cells. This study introduces a modular and tunable SNAr-based reactive group for targeting reactive cysteines in the human proteome and illustrates the pharmacological utility of this electrophilic series.
Sigma-Aldrich
4,6-Dimethyl-2-(methylsulfinyl)-3-nitropyridine