MilliporeSigma
Mechanistic insights on acrylate insertion polymerization.
Journal of the American Chemical Society (2010-03-09)
Damien Guironnet, Lucia Caporaso, Boris Neuwald, Inigo Göttker-Schnetmann, Luigi Cavallo, Stefan Mecking
ABSTRACT
Complexes [{(PwedgeO)PdMe}(n)] (1(n); PwedgeO = kappa(2)-P,O-Ar(2)PC(6)H(4)SO(2)O with Ar = 2-MeOC(6)H(4)) are a single-component precursor of the (PwedgeO)PdMe fragment devoid of additional coordinating ligands, which also promotes the catalytic oligomerization of acrylates. Exposure of 1(n) to methyl acrylate afforded the two diastereomeric chelate complexes [(PwedgeO)Pd{kappa(2)-C,O-CH(C(O)OMe)CH(2)CH(C(O)OMe)CH(2)CH(3)}] (3-meso and 3-rac) resulting from two consecutive 2,1-insertions of methyl acrylate into the Pd-Me bond with the same or opposite stereochemistry, respectively, in a 3:2 ratio as demonstrated by comprehensive NMR spectroscopic studies and single crystal X-ray diffraction. These six-membered chelate complexes are direct key models for intermediates of acrylate insertion polymerization, and also ethylene-acrylate copolymerization to high acrylate content copolymers. Studies of the binding of various substrates (pyridine, dmso, ethylene and methyl acrylate) to 3-meso and 3-rac show that hindered displacement of the chelating carbonyl moiety by pi-coordination of incoming monomer significantly retards, but does not prohibit, polymerization. For 3-meso,3-rac + C(2)H(4) right arrow over left arrow 3-meso-C(2)H(4,) 3-rac-C(2)H(4) an equilibrium constant K(353 K) approximately 2 x 10(-3) L mol(-1) was estimated. Reaction of 3-meso, 3-rac with methyl acrylate afforded higher insertion products [(PwedgeO)Pd(C(4)H(6)O(2))(n)Me] (n = 3, 4) as observed by electrospray ionization mass spectrometry (ESI-MS). Theoretical studies by DFT methods of consecutive acrylate insertion provide relative energies of intermediates and transition states, which are consistent with the aforementioned experimental observations, and give detailed insights to the pathways of multiple consecutive acrylate insertions. Acrylate insertion into 3-meso,3-rac is associated with an overall energy barrier of ca. 100 kJ mol(-1).