MilliporeSigma
  • NDST3 deacetylates α-tubulin and suppresses V-ATPase assembly and lysosomal acidification.
NDST3 deacetylates α-tubulin and suppresses V-ATPase assembly and lysosomal acidification.
The EMBO journal (2021-08-27)
Qing Tang, Mingming Liu, Yang Liu, Ran-Der Hwang, Tao Zhang, Jiou Wang
ABSTRACT
Lysosomes are key organelles maintaining cellular homeostasis in health and disease. Here, we report the identification of N-deacetylase and N-sulfotransferase 3 (NDST3) as a potent regulator of lysosomal functions through an unbiased genetic screen. NDST3 constitutes a new member of the histone deacetylase (HDAC) family and catalyzes the deacetylation of α-tubulin. Loss of NDST3 promotes assembly of the V-ATPase holoenzyme on the lysosomal membrane and thereby increases the acidification of the organelle. NDST3 is downregulated in tissues and cells from patients carrying the C9orf72 hexanucleotide repeat expansion linked to the neurodegenerative diseases amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD). Deficiency in C9orf72 decreases the level of NDST3, and downregulation of NDST3 exacerbates the proteotoxicity of poly-dipeptides generated from the C9orf72 hexanucleotide repeats. These results demonstrate a previously unknown regulatory mechanism through which microtubule acetylation regulates lysosomal activities and suggest that NDST3 could be targeted to modulate microtubule and lysosomal functions in relevant diseases.
Product Description
Millipore
PVDF Membrane Filter, 0.45 μm Pore Size, Durapore®, filter diam. 25 mm, hydrophobic
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder