MilliporeSigma
  • Total Synthesis of Emmyguyacins A and B, Potential Fusion Inhibitors of Influenza Virus.
Total Synthesis of Emmyguyacins A and B, Potential Fusion Inhibitors of Influenza Virus.
Organic letters (2018-10-24)
Santanu Jana, Vikram A Sarpe, Suvarn S Kulkarni
ABSTRACT
Fungal glycolipids emmyguyacins A and B inhibit the pH-dependent conformational change of hemaglutinin A during replication of the Influenza virus. Herein, we report the first total synthesis and structure confirmation of emmyguyacins A and B. Our efficient route, which involves regioselective functionalization of trehalose, allows rapid access to adequate amounts of chemically pure emmyguyacin analogues including the desoxylate derivatives for SAR studies.
Sigma-Aldrich
Methyl 11-bromoundecanoate, 95%