MilliporeSigma
  • Coelenterazine Analogue with Human Serum Albumin-Specific Bioluminescence.
Coelenterazine Analogue with Human Serum Albumin-Specific Bioluminescence.
Bioconjugate chemistry (2020-11-26)
Ryo Nishihara, Kazuki Niwa, Tatsunosuke Tomita, Ryoji Kurita
ABSTRACT
A synthetic luciferin comprising an imidazopyrazinone core, named HuLumino1, was designed to generate specific bioluminescence with human serum albumin (HSA) in real serum samples. HuLumino1 was developed by attaching a methoxy-terminated alkyl chain to C-6 of coelenterazine and by eliminating a benzyl group at C-8. HSA levels were quantified within 5% error margins of an enzyme-linked immunosorbent assay without the need for any sample pretreatments because of the high specificity of HuLumino1.
Sigma-Aldrich
Coelenterazine, A cell-permeable aequorin luminophore that acts as a very sensitive and specific chemiluminescence probe of the superoxide anion.