MilliporeSigma
  • Antitumor polycyclic acridines. 8.(1) Synthesis and telomerase-inhibitory activity of methylated pentacyclic acridinium salts.
Antitumor polycyclic acridines. 8.(1) Synthesis and telomerase-inhibitory activity of methylated pentacyclic acridinium salts.
Journal of medicinal chemistry (2002-01-25)
Robert A Heald, Chetna Modi, Jenny C Cookson, Ian Hutchinson, Charles A Laughton, Sharon M Gowan, Lloyd R Kelland, Malcolm F G Stevens
ABSTRACT
Two short routes to novel methylated pentacyclic quinoacridinium salts have been devised. New compounds display telomerase-inhibitory potency (<1 microM) in the TRAP assay. 3,11-Difluoro-6,8,13-trimethyl-8H-quino[4,3,2-kl]acridinium methosulfate (12d, RHPS4, NSC 714187) has a higher selectivity for triplex and quadruplex DNA structures than the 3,6,8,11,13-pentamethyl analogue (12c, RHPS3, NSC 714186) and a low overall growth-inhibitory activity in the NCI 60 cell panel (mean GI(50) 13.18 microM); in addition, the activity profile of 12d does not COMPARE with agents of the topoisomerase II class. Compound 12d is soluble in water, stable in the pH range of 5-9, efficiently transported into tumor cells, and is currently the lead structure for further elaboration in this new class of telomerase inhibitor.
Sigma-Aldrich
RHPS4 methosulfate, ≥98% (HPLC)