MilliporeSigma
  • Lighting up bioluminescence with coelenterazine: strategies and applications.
Lighting up bioluminescence with coelenterazine: strategies and applications.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2016-03-25)
Tianyu Jiang, Lupei Du, Minyong Li
ABSTRACT
Bioluminescence-based techniques, such as bioluminescence imaging, BRET and dual-luciferase reporter assay systems, have been widely used to examine a myriad of biological processes. Coelenterazine (CTZ), a luciferin or light-producing compound found in bioluminescent organisms, has sparked great curiosity and interest in searching for analogues with improved photochemical properties. This review summarizes the current development of coelenterazine analogues, their bioluminescence properties, and the rational design of caged coelenterazine towards biotargets, as well as their applications in bioassays. It should be emphasized that the design of caged luciferins can provide valuable insight into detailed molecular processes in organisms and will be a trend in the development of bioluminescent molecules.
Sigma-Aldrich
Coelenterazine, A cell-permeable aequorin luminophore that acts as a very sensitive and specific chemiluminescence probe of the superoxide anion.