MilliporeSigma
  • Intact protein separation by one- and two-dimensional liquid chromatography for the comparative proteomic separation of partitioned serum or plasma.
Intact protein separation by one- and two-dimensional liquid chromatography for the comparative proteomic separation of partitioned serum or plasma.
Methods in molecular biology (Clifton, N.J.) (2011-04-07)
Simon Sheng, Helena Skalnikova, Andrew Meng, John Tra, Qin Fu, Allen Everett, Jennifer Van Eyk
ABSTRACT
Serum- and plasma-based biomarker discovery requires technologies with specific capabilities: sufficient proteome coverage and depth, technical reproducibly, and the scalability to enable analysis on a large number of samples at reasonable cost. We have shown that plasma samples processed using IgY LC10 Proteome Partitioning kits to remove the most highly abundant proteins selectively, followed by intact protein separation by two-dimensional liquid chromatography (2DLC, chromatofocusing, and reversed phase) can uniquely enrich for middle to lower-abundant proteins. Equally, 1DLC (reversed phase) separation of intact proteins is complementary to 2DLC. The serial use of a single piece of equipment can be prohibitively time consuming and thus, this chapter also describes the harmonization of multiple LC instruments in order to minimize technical variation and ensure reproducibility. These technical improvements allow large numbers of individual clinical samples to be analyzed with multiple instruments in a timely manner, while retaining optimal reproducibility and allowing precise differential analysis at the proteome scale.
Product Description
Millipore
Seppro® Neutralization Buffer
Millipore
Seppro® Stripping Buffer
Sigma-Aldrich
Albumin from human serum, lyophilized powder, ≥97% (agarose gel electrophoresis)
Millipore
Seppro® Dilution buffer