MilliporeSigma
  • Improving the sensitivity of cellulose fiber-based lateral flow assay by incorporating a water-dissolvable polyvinyl alcohol dam.
Improving the sensitivity of cellulose fiber-based lateral flow assay by incorporating a water-dissolvable polyvinyl alcohol dam.
Cellulose (London, England) (2021-07-27)
Nur Alam, Li Tong, Zhibin He, Ruihua Tang, Laboni Ahsan, Yonghao Ni
ABSTRACT
Lateral flow assay (LFA) is an important point-of-care (POC) test platform due to the associated portability, on-site testing, and low cost for diagnosis of pathogen infections and disease biomarkers. However, compared to high-end analyzers in hospitals, LFA devices, in particular, paper- based LFA tests, fall short in accuracy. This study focuses on two ways to improve LFAs: (1) using cellulose fibers, rather than glass fibers for a sample pad, and (2) incorporating a one-step simple, facile, and low cost PVA dam into the LFA. Both strategies (cellulose fiber as a sample pad and water dissolvable PVA dam) contributed to delaying the controlled biomolecule's flow through the nitrocellulose membrane's capillary channels resulting in increased bio-recognition time, thus contributing to the enhancement of LFA sensitivity. PVA modified cellulose fiber-based LFA demonstrated 10 times higher sensitivity than the cellulose fiber-based unmodified LFA, whereas 2 times enhancement was obtained in the cellulose fiber-based sample pad LFA compared to the glass fiber-based sample pad LFA. Ultimately, 20 times increase in sensitivity was achieved in the modified LFA device. This study shows that PVA and eco-friendly cellulose fibers could be incorporated into other paper based POC testing devices for future development. The online version contains supplementary material available at 10.1007/s10570-021-04083-3.
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (HPLC), buffered aqueous solution
Sigma-Aldrich
Anti-Human IgG (whole molecule) antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgG (γ-chain specific)-Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution