MilliporeSigma
  • Suppression of tumor-associated neutrophils by lorlatinib attenuates pancreatic cancer growth and improves treatment with immune checkpoint blockade.
Suppression of tumor-associated neutrophils by lorlatinib attenuates pancreatic cancer growth and improves treatment with immune checkpoint blockade.
Nature communications (2021-06-09)
Sebastian R Nielsen, Jan E Strøbech, Edward R Horton, Rene Jackstadt, Anu Laitala, Marina C Bravo, Giorgia Maltese, Adina R D Jensen, Raphael Reuten, Maria Rafaeva, Saadia A Karim, Chang-Il Hwang, Luis Arnes, David A Tuveson, Owen J Sansom, Jennifer P Morton, Janine T Erler
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) patients have a 5-year survival rate of only 8% largely due to late diagnosis and insufficient therapeutic options. Neutrophils are among the most abundant immune cell type within the PDAC tumor microenvironment (TME), and are associated with a poor clinical prognosis. However, despite recent advances in understanding neutrophil biology in cancer, therapies targeting tumor-associated neutrophils are lacking. Here, we demonstrate, using pre-clinical mouse models of PDAC, that lorlatinib attenuates PDAC progression by suppressing neutrophil development and mobilization, and by modulating tumor-promoting neutrophil functions within the TME. When combined, lorlatinib also improves the response to anti-PD-1 blockade resulting in more activated CD8 + T cells in PDAC tumors. In summary, this study identifies an effect of lorlatinib in modulating tumor-associated neutrophils, and demonstrates the potential of lorlatinib to treat PDAC.
Sigma-Aldrich
Histopaque®-1119, sterile-filtered, density: 1.119
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)