MilliporeSigma
  • HBsAg Dampened STING Associated Activation of NK Cells in HBeAg-Negative CHB Patients.
HBsAg Dampened STING Associated Activation of NK Cells in HBeAg-Negative CHB Patients.
International journal of molecular sciences (2021-07-25)
Bingqing Zheng, Yating Yu, Zhaoyi Pan, Yujie Feng, Huajun Zhao, Qiuju Han, Jian Zhang
ABSTRACT
NK cells play crucial roles in defending against persistent HBV. However, NK cells present dysfunction in chronic hepatitis B virus (CHB) infection, and the associated mechanism is still not fully understood. Except for the regulatory receptors, NK cells could also be regulated by the surface and intracellular pattern recognition receptors (PRRs). In the present study, we found that the level of the adaptor of DNA sensor STING in NK cells was significantly decreased in HBeAg-negative CHB patients, and it was positively associated with the degranulation ability of NK cells. Compared to NK cells from healthy donors, NK cells from HBeAg-negative CHB patients displayed a lower responsiveness to cGAMP stimulation. Further investigation showed that HBsAg could inhibit the STING expression in NK cells and suppress the response of NK cells to cGAMP. Significantly, STAT3 was identified to be a transcription factor that directly regulated STING transcription by binding to the promoter. In addition, STAT3 positively regulated the STING associated IFN-α response of NK cells. These findings suggested that STING is an important adaptor in NK cell recognition and activation, while HBsAg disturbs NK cell function by the STAT3-STING axis, providing a new mechanism of NK disability in HBeAg-negative CHB infection.
Sigma-Aldrich
Magna ChIP® A/G Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads.