MilliporeSigma
MINSTED fluorescence localization and nanoscopy.
Nature photonics (2021-05-07)
Michael Weber, Marcel Leutenegger, Stefan Stoldt, Stefan Jakobs, Tiberiu S Mihaila, Alexey N Butkevich, Stefan W Hell
ABSTRACT
We introduce MINSTED, a fluorophore localization and super-resolution microscopy concept based on stimulated emission depletion (STED) that provides spatial precision and resolution down to the molecular scale. In MINSTED, the intensity minimum of the STED doughnut, and hence the point of minimal STED, serves as a movable reference coordinate for fluorophore localization. As the STED rate, the background and the required number of fluorescence detections are low compared with most other STED microscopy and localization methods, MINSTED entails substantially less fluorophore bleaching. In our implementation, 200-1,000 detections per fluorophore provide a localization precision of 1-3nm in standard deviation, which in conjunction with independent single fluorophore switching translates to a -100-fold improvement in far-field microscopy resolution over the diffraction limit. The performance of MINSTED nanoscopy is demonstrated by imaging the distribution of Mic60 proteins in the mitochondrial inner membrane of human cells.
Product Description
Roche
Streptavidin, recombinant, liquid, ≥17 units/mg protein, pH range 5.0-9.0
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O