MilliporeSigma
  • Nuclear localisation of 53BP1 is regulated by phosphorylation of the nuclear localisation signal.
Nuclear localisation of 53BP1 is regulated by phosphorylation of the nuclear localisation signal.
Biology of the cell (2018-04-01)
Patrick von Morgen, Tomas Lidak, Zuzana Horejsi, Libor Macurek
ABSTRACT
Repair of damaged DNA is essential for maintaining genomic stability. TP53-binding protein 1 (53BP1) plays an important role in repair of the DNA double-strand breaks. Nuclear localisation of 53BP1 depends on importin β and nucleoporin 153, but the type and location of 53BP1 nuclear localisation signal (NLS) have yet to be determined. Here, we show that nuclear import of 53BP1 depends on two basic regions, namely 1667-KRK-1669 and 1681-KRGRK-1685, which are both needed for importin binding. Lysine 1667 is essential for interaction with importin and its substitution to arginine reduced nuclear localisation of 53BP1. Furthermore, we have found that CDK1-dependent phosphorylation of 53BP1 at S1678 impairs importin binding during mitosis. Phosphorylation-mimicking mutant S1678D showed reduced nuclear localisation, suggesting that phosphorylation of the NLS interferes with nuclear import of the 53BP1 CONCLUSIONS: We show that 53BP1 contains a classical bipartite NLS 1666-GKRKLITSEEERSPAKRGRKS-1686, which enables the importin-mediated nuclear transport of 53BP1. Additionally, we found that posttranslational modification within the NLS region can regulate 53BP1 nuclear import. Our results indicate that integrity of the NLS is important for 53BP1 nuclear localisation. Precise mapping of the NLS will facilitate further studies on the effect of posttranslational modifications and somatic mutations on the nuclear localisation 53BP1 and DNA repair.
Sigma-Aldrich
Anti-Importin α antibody, Mouse monoclonal, clone IM-75, purified from hybridoma cell culture
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)