MilliporeSigma
  • F4, a collagen XIX-derived peptide, inhibits tumor angiogenesis through αvβ3 and α5β1 integrin interaction.
F4, a collagen XIX-derived peptide, inhibits tumor angiogenesis through αvβ3 and α5β1 integrin interaction.
Cell adhesion & migration (2021-07-27)
Jean-Baptiste Oudart, Matthieu Villemin, Bertrand Brassart, Christèle Sellier, Christine Terryn, Aurélie Dupont-Deshorgue, Jean Claude Monboisse, François-Xavier Maquart, Laurent Ramont, Sylvie Brassart-Pasco
ABSTRACT
We previously demonstrated that F4 peptide (CNPEDCLYPVSHAHQR) from collagen XIX was able to inhibit melanoma cell migrationin vitro and cancer progression in a mouse melanoma model. The aim of the present work was to study the anti-angiogenic properties of F4 peptide. We demonstrated that F4 peptide inhibited VEGF-induced pseudo-tube formation on Matrigel by endothelial cells and endothelial sprouting in a rat aortic ring assay. By affinity chromatography, we identified αvβ3 and α5β1 integrins as potential receptors for F4 peptide on endothelial cell surface. Using solid phase assays, we proved the direct interaction between F4 and both integrins. Taken together, our results demonstrate that F4 peptide is a potent antitumor agent inhibiting both angiogenesis and tumor cell migration.
Sigma-Aldrich
Anti-Integrin β1 Antibody, cytosolic, Chemicon®, from rabbit
Pricing and availability is not currently available.