MilliporeSigma
Films, Buckypapers and Fibers from Clay, Chitosan and Carbon Nanotubes.
Nanomaterials (Basel, Switzerland) (2011-04-06)
Thomas M Higgins, Holly Warren, Marc In Het Panhuis
ABSTRACT
The mechanical and electrical characteristics of films, buckypapers and fiber materials from combinations of clay, carbon nanotubes (CNTs) and chitosan are described. The rheological time-dependent characteristics of clay are maintained in clay-carbon nanotube-chitosan composite dispersions. It is demonstrated that the addition of chitosan improves their mechanical characteristics, but decreases electrical conductivity by three-orders of magnitude compared to clay-CNT materials. We show that the electrical response upon exposure to humid atmosphere is influenced by clay-chitosan interactions, i.e., the resistance of clay-CNT materials decreases, whereas that of clay-CNT-chitosan increases.
Sigma-Aldrich
Chitosan, high molecular weight