MilliporeSigma
  • Lack of AR in LepRb Cells Disrupts Ambulatory Activity and Neuroendocrine Axes in a Sex-Specific Manner in Mice.
Lack of AR in LepRb Cells Disrupts Ambulatory Activity and Neuroendocrine Axes in a Sex-Specific Manner in Mice.
Endocrinology (2020-07-02)
Alexandra L Cara, Martin G Myers, Carol F Elias
ABSTRACT
Disorders of androgen imbalance, such as hyperandrogenism in females or hypoandrogenism in males, increase risk of visceral adiposity, type 2 diabetes, and infertility. Androgens act upon androgen receptors (AR) which are expressed in many tissues. In the brain, AR are abundant in hypothalamic nuclei involved in regulation of reproduction and energy homeostasis, yet the role of androgens acting via AR in specific neuronal populations has not been fully elucidated. Leptin receptor (LepRb)-expressing neurons coexpress AR predominantly in hypothalamic arcuate and ventral premammillary nuclei (ARH and PMv, respectively), with low colocalization in other LepRb neuronal populations, and very low colocalization in the pituitary gland and gonads. Deletion of AR from LepRb-expressing cells (LepRbΔAR) has no effect on body weight, energy expenditure, and glucose homeostasis in male and female mice. However, LepRbΔAR female mice show increased body length later in life, whereas male LepRbΔAR mice show an increase in spontaneous ambulatory activity. LepRbΔAR mice display typical pubertal timing, estrous cycles, and fertility, but increased testosterone levels in males. Removal of sex steroid negative feedback action induced an exaggerated rise in luteinizing hormone in LepRbΔAR males and follicle-stimulating hormone in LepRbΔAR females. Our findings show that AR can directly affect a subset of ARH and PMv neurons in a sex-specific manner and demonstrate specific androgenic actions in the neuroendocrine hypothalamus.
Sigma-Aldrich
Anti-Estrogen Receptor α Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
REDExtract-N-Amp Tissue PCR Kit, sufficient for 10 reactions, sufficient for 100 reactions, sufficient for 1000 reactions, hotstart, dNTPs included