MilliporeSigma
  • Abraxas suppresses DNA end resection and limits break-induced replication by controlling SLX4/MUS81 chromatin loading in response to TOP1 inhibitor-induced DNA damage.
Abraxas suppresses DNA end resection and limits break-induced replication by controlling SLX4/MUS81 chromatin loading in response to TOP1 inhibitor-induced DNA damage.
Nature communications (2021-07-18)
Xiao Wu, Bin Wang
ABSTRACT
Although homologous recombination (HR) is indicated as a high-fidelity repair mechanism, break-induced replication (BIR), a subtype of HR, is a mutagenic mechanism that leads to chromosome rearrangements. It remains poorly understood how cells suppress mutagenic BIR. Trapping of Topoisomerase 1 by camptothecin (CPT) in a cleavage complex on the DNA can be transformed into single-ended double-strand breaks (seDSBs) upon DNA replication or colliding with transcriptional machinery. Here, we demonstrate a role of Abraxas in limiting seDSBs undergoing BIR-dependent mitotic DNA synthesis. Through counteracting K63-linked ubiquitin modification, Abraxas restricts SLX4/Mus81 recruitment to CPT damage sites for cleavage and subsequent resection processed by MRE11 endonuclease, CtIP, and DNA2/BLM. Uncontrolled SLX4/MUS81 loading and excessive end resection due to Abraxas-deficiency leads to increased mitotic DNA synthesis via RAD52- and POLD3- dependent, RAD51-independent BIR and extensive chromosome aberrations. Our work implicates Abraxas/BRCA1-A complex as a critical regulator that restrains BIR for protection of genome stability.
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
α-Amanitin, from Amanita phalloides, ≥85% (HPLC), powder
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Ubiquitin Antibody, Lys63-Specific, clone Apu3, rabbit monoclonal, clone Apu3, from rabbit
Sigma-Aldrich
Anti-RNF168 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Mirin, ≥98% (HPLC), powder
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
VE-821, ≥98% (HPLC)
Sigma-Aldrich
Anti-Lamin A (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Exo1, from rabbit
Sigma-Aldrich
Anti-Rad51 (Ab-1) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Hydroxyurea, 98%, powder