MilliporeSigma
  • Active Protection: Learning-Activated Raf/MAPK Activity Protects Labile Memory from Rac1-Independent Forgetting.
Active Protection: Learning-Activated Raf/MAPK Activity Protects Labile Memory from Rac1-Independent Forgetting.
Neuron (2018-03-20)
Xuchen Zhang, Qian Li, Lianzhang Wang, Zhong-Jian Liu, Yi Zhong
ABSTRACT
Active forgetting explains the intrinsic instability of a labile memory lasting for hours. However, how such memory maintains stability against unwanted disruption is not completely understood. Here, we report a learning-activated active protection mechanism that enables labile memory to resist disruptive sensory experiences in Drosophila. Aversive olfactory conditioning activates mitogen-activated protein kinase (MAPK) transiently in the mushroom-body γ lobe, where labile-aversive memory is stored. This increased MAPK activity significantly prolongs labile memory retention and enhances its resistance to disruption induced by heat shock, electric shock, or odor reactivation. Such experience-induced forgetting cannot be prevented by inhibition of Rac1 activity. Instead, protection of Rac1-independent forgetting correlates with non-muscle myosin II activity and persistence of learning-induced presynaptic structural changes. Increased Raf/MAPK activity, together with suppressed Rac1 activity, completely blocks labile memory decay. Thus, learning not only leads to memory formation, but also activates active protection and active forgetting to regulate the formed memory.
Sigma-Aldrich
3-Octanol, 99%