MilliporeSigma
  • Upregulation of erythropoietin and its receptor expression in the rat carotid body during chronic and intermittent hypoxia.
Upregulation of erythropoietin and its receptor expression in the rat carotid body during chronic and intermittent hypoxia.
Advances in experimental medicine and biology (2009-06-19)
S Y Lam, G L Tipoe, M L Fung
ABSTRACT
The carotid body (CB) plays important roles in cardiorespiratory changes in intermittent hypoxia (IH). Erythropoietin (EPO), a hypoxia-inducible factor (HIF)-1 target gene, is present in the chemoreceptive type-I cells in the CB but its expression and role in IH resembling sleep apnoeic conditions are not known. We hypothesized that IH upregulates the expression of EPO and its receptor (EPOr) in the rat CB. The CB expressions of EPO and EPOr were examined in rats breathing 10% O(2) (in isobaric chamber for CH, 24 hour/day) or in IH (cyclic between air and 5% O(2) per minute, 8 hour/day) for 3-28 days. Immunohistochemical studies revealed that the EPO and EPOr proteins were localized in CB glomic clusters. The proportional amount of cells with positive staining of EPO immunoreactivities was significantly increased in both IH and CH groups when compared with the normoxic control. The EPO expression was more markedly increased in the CH than that of the IH groups throughout the time course, reaching a peak level at day 14. The positive EPOr immunostaining was increased significantly in the 3-day CH group. By day 14, the EPOr expression elevated considerably at peak levels in both IH and CH rats, whereas the elevation was greater in the CH rats. These results suggest an upregulation of EPO and its receptor expression in the rat CB under IH and CH conditions, presumably mediated by the activation of HIF-1 pathway. The increased EPO binding to its receptor might play a role in the enhancement of CB excitability during the early pathogenesis in patients with sleep-disordered breathing.
Sigma-Aldrich
Anti-Erythropoietin Receptor antibody produced in goat, affinity isolated antibody