MilliporeSigma
  • Cell surface engineering of renal cell carcinoma with glycosylphosphatidylinositol-anchored TIMP-1 blocks TGF- β 1 activation and reduces regulatory ID gene expression.
Cell surface engineering of renal cell carcinoma with glycosylphosphatidylinositol-anchored TIMP-1 blocks TGF- β 1 activation and reduces regulatory ID gene expression.
Biological chemistry (2013-05-15)
Susan Notohamiprodjo, Roghieh Djafarzadeh, Nicole Rieth, Monika Hofstetter, Carsten Jaeckel, Peter J Nelson
ABSTRACT
Tissue inhibitor of metalloproteinase 1 (TIMP-1) controls matrix metalloproteinase activity through 1:1stoichiometric binding. Human TIMP-1 fused to a glycosylphosphatidylinositol(GPI) anchor (TIMP-1 - GPI) shifts the activity of TIMP-1 from the extracellular matrix to the cell surface. TIMP-1 - GPI treated renal cell carcinoma cells show increased apoptosis and reduced proliferation.Transcriptomic profiling and regulatory pathway mapping were used to identify the potential mechanisms driving these effects. Significant changes in the DNA binding inhibitors, TGF- β 1/SMAD and BMP pathways resulted from TIMP-1 - GPI treatment. These events were linked to reduced TGF- β 1 signaling mediated by inhibition of proteolytic processing of latent TGF- β 1 by TIMP-1 - GPI.
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-MMP-1 (Ab-1) Mouse mAb (41-1E5), liquid, clone 41-1E5, Calbiochem®
Sigma-Aldrich
Anti-TIMP-1 (Ab-1) Mouse mAb (7-6C1), liquid, clone 7-6C1, Calbiochem®
Sigma-Aldrich
Anti-MMP-2 (Ab-3) Mouse mAb (42-5D11), liquid, clone 42-5D11, Calbiochem®