MilliporeSigma
Caspase-Dependent Cleavage of DDX21 Suppresses Host Innate Immunity.
mBio (2021-06-15)
Wei Wu, Yang Qu, Shengqing Yu, Sa Wang, Yuncong Yin, Qinfang Liu, Chunchun Meng, Ying Liao, Zaib Ur Rehman, Lei Tan, Cuiping Song, Xusheng Qiu, Weiwei Liu, Chan Ding, Yingjie Sun
ABSTRACT
DEAD (Glu-Asp-Ala-Glu) box RNA helicases have been proven to contribute to antiviral innate immunity. The DDX21 RNA helicase was identified as a nuclear protein involved in rRNA processing and RNA unwinding. DDX21 was also proven to be the scaffold protein in the complex of DDX1-DDX21-DHX36, which senses double-strand RNA and initiates downstream innate immunity. Here, we identified that DDX21 undergoes caspase-dependent cleavage after virus infection and treatment with RNA/DNA ligands, especially for RNA virus and ligands. Caspase-3/6 cleaves DDX21 at D126 and promotes its translocation from the nucleus to the cytoplasm in response to virus infection. The cytoplasmic cleaved DDX21 negatively regulates the interferon beta (IFN-β) signaling pathway by suppressing the formation of the DDX1-DDX21-DHX36 complex. Thus, our data identify DDX21 as a regulator of immune balance and most importantly uncover a potential role of DDX21 cleavage in the innate immune response to virus. IMPORTANCE Innate immunity serves as the first barrier against virus infection. DEAD (Glu-Asp-Ala-Glu) box RNA helicases, originally considered to be involved in RNA processing and RNA unwinding, have been shown to play an important role in antiviral innate immunity. The precise regulation of innate immunity is critical for the host because the aberrant production of cytokines leads to unexpected pathological consequences. Here, we identified that DDX21 was cleaved at D126 by virus infection and treatment with RNA/DNA ligands via the caspase-3/6-dependent pathway. The cytoplasmic cleaved DDX21 negatively regulates the IFN-β signaling pathway by suppressing the formation of the DDX1-DDX21-DHX36 complex. In sum, our data identify DDX21 as a regulator of immune balance and most importantly uncover a potential role of DDX21 cleavage in the innate immune response to virus.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture