MilliporeSigma
  • Absence of sterols constrains food quality of cyanobacteria for an invasive freshwater bivalve.
Absence of sterols constrains food quality of cyanobacteria for an invasive freshwater bivalve.
Oecologia (2012-03-09)
Timo Basen, Karl-Otto Rothhaupt, Dominik Martin-Creuzburg
ABSTRACT
The accumulation of cyanobacterial biomass may severely affect the performance of aquatic consumers. Here, we investigated the role of sterols in determining the food quality of cyanobacteria for the invasive clam Corbicula fluminea, which has become a common benthic invertebrate in many freshwater ecosystems throughout the world. In standardized growth experiments, juvenile clams were fed mixtures of different cyanobacteria (Anabaena variabilis, Aphanothece clathrata, Synechococcus elongatus) or sterol-containing eukaryotic algae (Cryptomonas sp., Nannochloropsis limnetica, Scenedesmus obliquus). In addition, the cyanobacterial food was supplemented with different sterols. We provide evidence that somatic growth of C. fluminea on cyanobacterial diets is constrained by the absence of sterols, as indicated by a growth-enhancing effect of sterol supplementation. Thus, our findings contribute to our understanding of the consequences of cyanobacterial mass developments for benthic consumers and highlight the importance of considering sterols as potentially limiting nutrients in aquatic food webs.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Stigmasterol, ~95%