MilliporeSigma
  • Nitration of tyrosine 10 critically enhances amyloid β aggregation and plaque formation.
Nitration of tyrosine 10 critically enhances amyloid β aggregation and plaque formation.
Neuron (2011-09-10)
Markus P Kummer, Michael Hermes, Andrea Delekarte, Thea Hammerschmidt, Sathish Kumar, Dick Terwel, Jochen Walter, Hans-Christian Pape, Simone König, Sigrun Roeber, Frank Jessen, Thomas Klockgether, Martin Korte, Michael T Heneka
ABSTRACT
Part of the inflammatory response in Alzheimer's disease (AD) is the upregulation of the inducible nitric oxide synthase (NOS2) resulting in increased NO production. NO contributes to cell signaling by inducing posttranslational protein modifications. Under pathological conditions there is a shift from the signal transducing actions to the formation of protein tyrosine nitration by secondary products like peroxynitrite and nitrogen dioxide. We identified amyloid β (Aβ) as an NO target, which is nitrated at tyrosine 10 (3NTyr(10)-Aβ). Nitration of Aβ accelerated its aggregation and was detected in the core of Aβ plaques of APP/PS1 mice and AD brains. NOS2 deficiency or oral treatment with the NOS2 inhibitor L-NIL strongly decreased 3NTyr(10)-Aβ, overall Aβ deposition and cognitive dysfunction in APP/PS1 mice. Further, injection of 3NTyr(10)-Aβ into the brain of young APP/PS1 mice induced β-amyloidosis. This suggests a disease modifying role for NOS2 in AD and therefore represents a potential therapeutic target.
Sigma-Aldrich
Anti-IDE, N-Terminal (97-273) Rabbit pAb, liquid, Calbiochem®