MilliporeSigma
  • A NOVEL NOX/PHOX-CD38-NAADP-TFEB AXIS IMPORTANT FOR MACROPHAGE ACTIVATION DURING BACTERIAL PHAGOCYTOSIS.
A NOVEL NOX/PHOX-CD38-NAADP-TFEB AXIS IMPORTANT FOR MACROPHAGE ACTIVATION DURING BACTERIAL PHAGOCYTOSIS.
Autophagy (2021-04-06)
Mehran Najibi, Havisha H Honwad, Joseph A Moreau, Stephanie M Becker, Javier E Irazoqui
ABSTRACT
Macrophage activation in the presence of bacterial cells and molecules entails complex programs of gene expression. How such triggers elicit specific gene expression programs is incompletely understood. We previously discovered that TFEB (transcription factor EB) is a key contributor to macrophage activation during bacterial phagocytosis. However, the mechanism linking phagocytosis of bacterial cells to TFEB activation and downstream pro-inflammatory cytokine induction remained unknown. We found that macrophages lacking both TFEB and TFE3 (transcription factor E3) were unable to mount a pro-inflammatory phenotype in response to bacterial infection. The NOX/PHOX (NADPH oxidase)-dependent oxidative burst was required for nuclear translocation of TFEB during phagocytosis of Gram-positive or -negative bacteria, and reactive oxygen species (ROS) were sufficient to trigger TFEB activation in a CD38- and NAADP (nicotinic acid adenine dinucleotide phosphate)-dependent manner. Consistent with the Ca2+-releasing activity of NAADP, intracellular Ca2+ chelation and PPP3/calcineurin inhibition prevented TFEB activation by phagocytosis and ROS (reactive oxygen species), impairing the induction of pro-inflammatory cytokines such as IL6 and TNF/TNFα. Therefore, here we describe a previously unknown pathway that links phagocytosis with macrophage pro-inflammatory polarization via TFEB and related transcription factor TFE3. These findings reveal that activation of TFEB and TFE3 is a key regulatory event for the activation of macrophages, and have important implications for infections, inflammation, cancer, obesity, and atherosclerosis.
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
ML-SA1, ≥95% (HPLC)