MilliporeSigma
  • Biologically active arborinane-type triterpenoids and anthraquinones from Rubia yunnanensis.
Biologically active arborinane-type triterpenoids and anthraquinones from Rubia yunnanensis.
Journal of natural products (2011-10-07)
Jun-Ting Fan, Bin Kuang, Guang-Zhi Zeng, Si-Meng Zhao, Chang-Jiu Ji, Yu-Mei Zhang, Ning-Hua Tan
ABSTRACT
Twelve new arborinane-type triterpenoids (1-12) and four new anthraquinones (13-16), together with 50 known compounds, were isolated from the roots of Rubia yunnanensis. The structures of 1-16 were elucidated by spectroscopic data analysis and chemical methods. All compounds were evaluated for their cytotoxic, antibacterial, and antifungal activities. Rubiyunnanol C (5) is the first example of an arborinane-type triterpenoid with a double bond at C-8-C-9.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
(±)-Miconazole nitrate salt, imidazole antibiotic
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder