MilliporeSigma
  • Modeling Human TBX5 Haploinsufficiency Predicts Regulatory Networks for Congenital Heart Disease.
Modeling Human TBX5 Haploinsufficiency Predicts Regulatory Networks for Congenital Heart Disease.
Developmental cell (2020-12-16)
Irfan S Kathiriya, Kavitha S Rao, Giovanni Iacono, W Patrick Devine, Andrew P Blair, Swetansu K Hota, Michael H Lai, Bayardo I Garay, Reuben Thomas, Henry Z Gong, Lauren K Wasson, Piyush Goyal, Tatyana Sukonnik, Kevin M Hu, Gunes A Akgun, Laure D Bernard, Brynn N Akerberg, Fei Gu, Kai Li, Matthew L Speir, Maximilian Haeussler, William T Pu, Joshua M Stuart, Christine E Seidman, J G Seidman, Holger Heyn, Benoit G Bruneau
ABSTRACT
Haploinsufficiency of transcriptional regulators causes human congenital heart disease (CHD); however, the underlying CHD gene regulatory network (GRN) imbalances are unknown. Here, we define transcriptional consequences of reduced dosage of the CHD transcription factor, TBX5, in individual cells during cardiomyocyte differentiation from human induced pluripotent stem cells (iPSCs). We discovered highly sensitive dysregulation of TBX5-dependent pathways-including lineage decisions and genes associated with heart development, cardiomyocyte function, and CHD genetics-in discrete subpopulations of cardiomyocytes. Spatial transcriptomic mapping revealed chamber-restricted expression for many TBX5-sensitive transcripts. GRN analysis indicated that cardiac network stability, including vulnerable CHD-linked nodes, is sensitive to TBX5 dosage. A GRN-predicted genetic interaction between Tbx5 and Mef2c, manifesting as ventricular septation defects, was validated in mice. These results demonstrate exquisite and diverse sensitivity to TBX5 dosage in heterogeneous subsets of iPSC-derived cardiomyocytes and predicts candidate GRNs for human CHDs, with implications for quantitative transcriptional regulation in disease.
Sigma-Aldrich
Normal Rabbit IgG, This Normal Rabbit IgG is validated for use as a negative control in parallel with specific primary antibodies in ELISA, FC, Immunoblotting, IF, IHC, IP.
Sigma-Aldrich
Anti-TBX5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution