MilliporeSigma
  • NO releases bombesin-like immunoreactivity from enteric synaptosomes by cross-activation of protein kinase A.
NO releases bombesin-like immunoreactivity from enteric synaptosomes by cross-activation of protein kinase A.
The American journal of physiology (1999-06-11)
M Kurjak, R Fritsch, D Saur, V Schusdziarra, H D Allescher
ABSTRACT
The effect of nitric oxide (NO) on the release of bombesin-like immunoreactivity (BLI) was examined in synaptosomes of rat small intestine. The NO donor S-nitroso-N-acetylpenicillamine (SNAP; 10(-7) to 10(-4) M) significantly stimulated BLI release. In the presence of the NO scavenger oxyhemoglobin (10(-3) M) or the guanylate cyclase inhibitor ODQ (10(-5) M), SNAP-induced BLI release was antagonized. In addition, SNAP increased the synaptosomal cGMP content and elevation of cGMP levels by zaprinast (3 x 10(-5) M), an inhibitor of the cGMP-specific phosphodiesterase (PDE) type 5, and increased basal and SNAP-induced BLI release. NO-induced BLI release was blocked by Rp-adenosine 3',5'-cyclic monophosphorothioate (3 x 10(-5) M and 10(-4) M), an inhibitor of the cAMP-dependent protein kinase A, whereas KT-5823 (3 x 10(-6) M) and Rp-8-(4-chlorophenylthio)-cGMP (5 x 10(-5) M), inhibitors of the cGMP-dependent protein kinase G, had no effect. Because cGMP inhibits the cAMP-specific PDE3, thereby increasing cAMP levels, the role of PDE3 was investigated. Trequinsin (10(-8) M), a specific blocker of PDE3, stimulated basal BLI release but had no additive effect on NO-induced release, suggesting a similar mechanism of action. These data demonstrate that because of a cross-activation of cAMP-dependent protein kinase A by endogenous cGMP BLI can be released by NO from enteric synaptosomes.
Sigma-Aldrich
Trequinsin hydrochloride, ≥98%