MilliporeSigma
Benzodihydrofurans from Cyperus teneriffae.
Journal of natural products (2011-04-21)
Ángel Amesty, Eleuterio Burgueño-Tapia, Pedro Joseph-Nathan, Ángel G Ravelo, Ana Estévez-Braun
ABSTRACT
Three new benzodihydrofurans (1-3) and seven known aromatic compounds (4-10) were isolated from the roots of Cyperus teneriffae. Vibrational circular dichroism spectroscopy was used to define the absolute configuration of 1.
Sigma-Aldrich
Eriodictyol, ≥95.0% (HPLC)