MilliporeSigma
  • Peptide Bond Formation of Amino Acids by Transient Masking with Silylating Reagents.
Peptide Bond Formation of Amino Acids by Transient Masking with Silylating Reagents.
Journal of the American Chemical Society (2021-05-01)
Wataru Muramatsu, Hisashi Yamamoto
ABSTRACT
A one-pot peptide bond-forming reaction has been developed using unprotected amino acids and peptides. Two different silylating reagents, HSi[OCH(CF3)2]3 and MTBSTFA, are instrumental for the successful implementation of this approach, being used for the activation and transient masking of unprotected amino acids and peptides at C-termini and N-termini, respectively. Furthermore, CsF and imidazole are used as catalysts, activating HSi[OCH(CF3)2]3 and also accelerating chemoselective silylation. This method is versatile as it tolerates side chains that bear a range of functional groups, while providing up to >99% yields of corresponding peptides without any racemization or polymerization.
Sigma-Aldrich
Tris[(1,1,1,3,3,3-hexafluoropropan-2-yl)oxy]silane