MilliporeSigma
  • Metabolite Identification and Pharmacokinetic Profiling of Isoflavones from Black Soybean in Rats Using Ultrahigh-Performance Liquid Chromatography with Linear-Ion-Trap-Orbitrap and Triple-Quadrupole Tandem Mass Spectrometry.
Metabolite Identification and Pharmacokinetic Profiling of Isoflavones from Black Soybean in Rats Using Ultrahigh-Performance Liquid Chromatography with Linear-Ion-Trap-Orbitrap and Triple-Quadrupole Tandem Mass Spectrometry.
Journal of agricultural and food chemistry (2018-11-18)
Juping Zhang, Qianxiang Guo, Meijin Wei, Junqi Bai, Juan Huang, Yuhong Liu, Ziren Su, Xiaohui Qiu
ABSTRACT
Black soybeans are rich in isoflavones, which have several beneficial health effects. In this study, a validated method based on UHPLC-MS/MS was developed to screen black-soybean metabolites in rat urine, bile, and plasma and to quantify the compounds (daidzein, genistein, glycitein, and daidzin) and their metabolites (daidzein-4'-β-d-glucuronide, genistein-7-β-d-glucuronide, and genistein-4'-β-d-glucuronide) in plasma. Thirty-seven compounds were tentatively detected in the biological samples. The method was fully validated in quantitative experiments, including in assessments of linearity (2.5-100 ng/mL for daidzein, genistein, and glycitein; 10-100 ng/mL for daidzin; 5-3125 ng/mL for genistein-7-β-d-glucuronide; and 5-1562.5 ng/mL for daidzein-4'-β-d-glucuronide and genistein-4'-β-d-glucuronide), matrix effects (85-115%), recovery (80-105%), precision (<10%), and accuracy (<10%). The compounds were stable throughout sample storage, treatment, and analysis. The method was first applied to detect IFs and metabolites in rats after oral administration of black-soybean extract. These results support the potential of this method for successful application in pharmacokinetic studies.
Sigma-Aldrich
Daidzein glucuronide potassium salt