MilliporeSigma
Neural Stem Cells Direct Axon Guidance via Their Radial Fiber Scaffold.
Neuron (2020-07-25)
Navjot Kaur, Wenqi Han, Zhuo Li, M Pilar Madrigal, Sungbo Shim, Sirisha Pochareddy, Forrest O Gulden, Mingfeng Li, Xuming Xu, Xiaojun Xing, Yutaka Takeo, Zhen Li, Kangrong Lu, Yuka Imamura Kawasawa, Begoña Ballester-Lurbe, Juan Antonio Moreno-Bravo, Alain Chédotal, José Terrado, Ignacio Pérez-Roger, Anthony J Koleske, Nenad Sestan
ABSTRACT
Neural stem cells directly or indirectly generate all neurons and macroglial cells and guide migrating neurons by using a palisade-like scaffold made of their radial fibers. Here, we describe an unexpected role for the radial fiber scaffold in directing corticospinal and other axons at the junction between the striatum and globus pallidus. The maintenance of this scaffold, and consequently axon pathfinding, is dependent on the expression of an atypical RHO-GTPase, RND3/RHOE, together with its binding partner ARHGAP35/P190A, a RHO GTPase-activating protein, in the radial glia-like neural stem cells within the ventricular zone of the medial ganglionic eminence. This role is independent of RND3 and ARHGAP35 expression in corticospinal neurons, where they regulate dendritic spine formation, axon elongation, and pontine midline crossing in a FEZF2-dependent manner. The prevalence of neural stem cell scaffolds and their expression of RND3 and ARHGAP35 suggests that these observations might be broadly relevant for axon guidance and neural circuit formation.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Roche
DIG RNA Labeling Mix, sufficient for 20 reactions, solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Neural Cell Adhesion Molecule L1 Antibody, clone 324, clone 324, Chemicon®, from rat
Sigma-Aldrich
Anti-Brain lipid binding protein Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
D-Lysine, ≥98% (HPLC)
Sigma-Aldrich
BCIP®/NBT Liquid Substrate System, ready to use solution
Sigma-Aldrich
Benzyl ether, 98%
Sigma-Aldrich
Cytochrome c Oxidase from bovine heart, 5 mg protein/mL
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Roche
RNase, from bovine pancreas
Sigma-Aldrich
Anti-RhoE/Rnd3 Antibody, clone 4, ascites fluid, clone 4, Upstate®