MilliporeSigma
  • The relationship between chronic exposure to arsenic through drinking water and hearing function in exposed population aged 10-49 years: A cross-sectional study.
The relationship between chronic exposure to arsenic through drinking water and hearing function in exposed population aged 10-49 years: A cross-sectional study.
Ecotoxicology and environmental safety (2021-01-22)
Reza Shokoohi, Mohammad Khazaei, Manoochehr Karami, Abdolmotaleb Seidmohammadi, Nima Berijani, Hassan Khotanlou, Zahra Torkshavand
ABSTRACT
It has been documented that arsenic has a potential risk to human health and identified as a risk factor for hearing impairment. However, there are few studies that confirm the ototoxic effect of arsenic, especially on the human auditory system. Therefore, the current study was conducted to investigate the correlation between auditory thresholds at different frequencies (0.25, 0.5, 1, 2, 4 and 8 kHz) and arsenic levels in drinking water samples. A total of 240 people, divided into two equal groups: exposed and reference, were selected for the auditory tests. It should be noted that, at frequencies from 0.25 to 1 kHz, no hearing loss was observed in the both groups. Based on the results, no significant correlations (p > 0.05) were found between hearing thresholds and confounding variables including gender and BMI. However, smoking and age are known to be the main variables for hearing loss in univariate regression analysis. In the case of age, the hearing loss risk in the older participants was increased compared with the younger participants (4 kHz (OR =1.09; 95% CI: 1.04, 1.13) and 8 kHz (OR =1.12; 95% CI: 1.06, 1.18)). Smoking habits had significant associations with hearing loss risk at 4 kHz (OR = 3.48; 95% CI: 1.47, 8.22) and 8 kHz (OR = 3.01; 95% CI: 1.14, 7.95). The multivariate regression analysis showed that age, smoking status, and exposure to arsenic were significantly associated with increased risk of hearing loss. Moreover, no statistically significant correlation (p˃0.05) was observed between arsenic exposure and hearing loss in the logistic regression model compared to the reference group. These outcomes suggest that further investigation and cohort studies with a larger number of participants should be conducted to find an association between arsenic exposure and hearing loss in general population.
Product Description
Supelco
Multielement standard solution 4 for ICP, TraceCERT®, in 10% nitric acid